Universal Inlet Adapters, Chemglass
Supplier: Chemglass
CG-1043-01 CG-1043-04 CG-1043-05 CG-1043-06 CG-1043-11 CG-1043-12 CG-1043-13 CG-1043-14
80065-986EA 111.43 CAD
80065-986 80065-988 80065-990 80065-992 80065-994 80065-996 80065-998 80066-000
Universal Inlet Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters have the listed size of Chem-Thread at the top for a vacuum tight seal on a variety of tubing diameters.
  • Will accept glass, metal tubes with diameters between 7 and 18 mm O.D.
Will accept glass, metal, or heavy wall plastic tubes such as thermometer wells, gas dispersion tubes, plain stem funnels and more.
Delivery information: Adapters are supplied complete with a Celcon compression cap and Viton O-Ring.