Thermocouple Adapters, Chemglass
Supplier: Chemglass
CG-1042-E-01 CG-1042-E-02 CG-1042-E-06 CG-1042-E-07 CG-1042-E-10
16002-624EA 159.57 CAD
16002-624 16002-626 16002-628 16002-630 16002-634
Thermocouple Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters feature a Chem-Thread at the top to make a vacuum tight seal.
  • Accepts 1/8" or 1/4" O.D. thermocouple probes
Lower portion of the adapter is reduced down to hold the thermocouple securely in place and away from the agitators.
Delivery information: Supplied complete with a Celcon cap and Viton O-Ring. Perfluoro O-Rings are also available.