Straight Inlet Adapters, [ST] Inner Joint, Chemglass
Supplier: Chemglass
CG-1012-C-05 CG-1012-C-01 CG-1012-C-02
16002-590EA 164.74 CAD
16002-590 16002-586 16002-588
Straight Inlet Adapters, [ST] Inner Joint, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters allow PTFE tubing to be connected to appartus for introduction of vacuum or pressure.
  • Designed for use with:
  • 1/2" TFE compression fitting
  • PFA compression fitting
  • Polypropylene compression fitting
The 1/2" O.D. ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.