Inlet Adapters, [ST] Joint, 90° Bend, Chemglass
Supplier: Chemglass
CG-1014-C-01 CG-1014-C-02 CG-1014-C-03
16002-594EA 118.58 CAD
16002-594 16002-596 16002-598
Inlet Adapters, [ST] Joint, 90° Bend, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters allow PTFE tubing to be connected to apparatus for introduction of vacuum or pressure.
  • Designed for use with:
  • 1/2" TFE compression fitting
  • PFA compression fitting
  • Polypropylene compression fitting
The 1/2" O.D. ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.