Inlet Adapters, [ST] Inner Joint, 90° Bend, Chemglass
Supplier: Chemglass
CG-1014-19 CG-1014-29
80065-722EA 82.06 CAD
80065-722 80065-724
Inlet Adapters, [ST] Inner Joint, 90° Bend, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters allow for flexible tubing to be connected to apparatus for introduction of vacuum or pressure.
Hose connection has an O.D. of 10 mm at the largest serration and is bent downward at a 90° angle to the inner joint.