Full Length Outer Ground Joints, Chemglass
Supplier: Chemglass
CG-101-01 CG-101-02 CG-101-03 CG-101-04 CG-101-05 CG-101-06 CG-101-07 CG-101-08 CG-101-09 CG-101-10 CG-101-11 CG-101-12 CG-101-13 CG-101-14 CG-101-15
80072-734EA 35.51 CAD
80072-734 80072-736 80072-738 80072-740 80072-742 80072-744 80072-746 80072-748 80072-750 80072-752 80072-754 80072-756 80072-758 80072-760 80072-762
Full Length Outer Ground Joints, Chemglass
Adapters Adapters, Conical Ground Joints
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below the joint.