Female Luer Lock Vacuum Adapters, Chemglass
Supplier: Chemglass
CG-1049-F-14 CG-1049-F-24
89094-918EA 387.36 CAD
89094-918 89094-920
Female Luer Lock Vacuum Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters have a 1/4"–28 thread that accepts a female luer lock adapter. This provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes, or syringes with a male luer lock fitting to a standard male taper inner joint.
  • Hose connection is 10 mm O.D. at the largest serration
The adapter enables easier sample concentration and clean-up while reducing the chances of cross-contamination.
Delivery information: Supplied complete with one 1/4"–28 Female to Female luer lock adapter.