98.0-100.5% (dry basis) USP, ACS
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
di-Potassium hydrogen orthophosphate, Dipotassium hydrogenphosphate, Potassium hydrogen phosphate, Dipotassium hydrogen phosphate, Dipotassium Phosphate, Dibasic Potassium Phosphate, Potassium Phosphate, Dibasic, (dipotassium hydrogen phosphate), Potassium phosphate dibasic, Dipotassium phosphate
Management of Change (MOC) category = R
BAKR3222-07
BAKR3222-65
CAJT3222-07CS
1471.91
CAD
CAJT3222-07
CAJT3222-65
98.0-100.5% (dry basis) USP, ACS
di-Potassium hydrogen phosphate
Formula:
K₂HPO₄ MW: 174.18 g/mol Melting Pt: 340 |
MDL Number:
MFCD00011383 CAS Number: 7758-11-4 Merck Index: 13,07743 |
|
Specification Test Results
Meets U.S.P Requirements | |
GMP Manufactured Product | |
Meets ACS Reagent Chemical Requirements | |
CAUTION: For Manufacturing, processing or repackaging | |
Bulk Pharmaceutical Chemical | |
USP - Assay (K₂HPO₄) (dried basis) | 98.0 - 100.5 % |
USP - Arsenic (As) | ≤ 3 ppm |
USP - Carbonate (CO₃) | Passes Test |
USP - Chloride (Cl) | ≤ 0.03 % |
USP - Fluoride (F) | ≤ 0.001 % |
USP - Identification A (USP) | Passes Test |
USP - Identification B (USP) | Passes Test |
USP - Insoluble Substances | ≤ 0.2 % |
USP - Iron (Fe) | ≤ 0.003 % |
USP - Loss on Drying | ≤ 1.0 % |
USP - Mono- or Tri- Basic Salt | Passes Test |
USP - pH (1 in 20) | 8.5 - 9.6 |
USP - Sodium (Na) | Passes Test |
USP - Sulfate (SO₄) | ≤ 0.1 % |
Assay (K₂HPO₄) (by acidimetry) | ≥ 99.0 % |
Chloride (Cl) | ≤ 0.003 % |
Fluoride (F) | ≤ 0.001 % |
Trace Impurities - ACS - Heavy Metals (as Pb) | ≤ 5 ppm |
Insoluble Matter | ≤ 0.01 % |
Trace Impurities - Iron (Fe) | ≤ 0.001 % |
Loss on Drying at 105°C | ≤ 1.0 % |
Nitrogen Compounds (as N) | ≤ 0.001 % |
pH of 5% Solution at 25°C | 8.5 - 9.6 |
Sodium (Na) | ≤ 0.05 % |
Sulfate (SO₄) | ≤ 0.005 % |
Trace Impurities - Arsenic (As) | ≤ 1 ppm |
Trace Impurities - Lead (Pb) | ≤ 5 ppm |
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. | |
Preserve in well closed container. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...