Distillation Adapters, with Tooled Rubber Fitting, Chemglass
Supplier: Chemglass
CG-1026-01 CG-1026-14 CG-1026-19
80065-852EA 182.62 CAD
80065-852 80065-854 80065-856
Distillation Adapters, with Tooled Rubber Fitting, Chemglass
Distillation Components Distillation Adapters
Adapters have a tubulation at the top for use with rubber fitting.
Fitting permits insertion of plain stem, partial immersion thermometers. With the same size inner joints on the bottom and sidearm at a 75° angle to one another.
Delivery information: Supplied with one rubber fitting.