tetra-Chloroauric(III) acid trihydrate, BAKER ANALYZED® ACS
Supplier: VWR International
|
Danger
|
Synonyms:
Gold(III) chloride trihydrate, Gold Chloride hydrate, Hydrogen tetrachloroaurate(III) trihydrate, Hydrogen tetrachloroaurate (III) trihydrate, tetra-Chloroauric acid trihydrate
CAJT2146-0
CAJT2146-3
BAKR2146-01
CAJT2146-0EA
20105.44
CAD
CAJT2146-0
CAJT2146-3
CAJT2146-01
tetra-Chloroauric(III) acid trihydrate, BAKER ANALYZED® ACS
tetra-Chloroauric (III) acid trihydrate
Formula:
HAuCl₄·3H₂O MW: 393.83 g/mol Melting Pt: 30 Density: 3.90 g/cm³ |
MDL Number:
MFCD00149904 CAS Number: 16961-25-4 UN: 3260 ADR: 8 II |
Specification Test Results
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | |
Material is very hygroscopic. Store in a cool place. | |
Assay (as Au) | ≥49.0 % |
Insoluble in Ether | ≤0.1 % |
Alkalies and Other Metals (as SO₄) | ≤0.2 % |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...