Hydrochloric acid 5.0 N, Biotech Reagent, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

Management of Change (MOC) category = R

0319-07 BAKR0319-22 BAKR0319-07
JT0319-07EA 2410.34 CAD
JT0319-07 CAJT0319-22 CAJT0319-07
Hydrochloric acid 5.0 N, Biotech Reagent, J.T.Baker®
Hydrochloric acid

Made from USP Purified Water and Hydrochloric Acid, NF (Multi-Compendial) which meets BP, Ph. Eur. and JP Chemical Specifications.


Filtered through a 0.2 micron filter.

Formula: HCl
Melting Pt: 48 °C
Density: 1.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-867-5
UN: 1789
ADR: 8,II
Merck Index: 12,08844

Order Now

Specification Test Results

GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Appearance (Clear, colorless liquid) Passes Test
Normality, eq/L 4.98 - 5.02
Sulfate (SO₄) ≤1 ppm
Heavy Metals (as Pb) ≤0.5 ppm
Iron (Fe) ≤0.1 ppm
Ammonium (NH₄) ≤3 ppm
Made from U.S.P. Purified Water and Hydrochloric Acid, N.F.
(Multi-Compendial) which meets BP,Ph.Eur.and JP Chemical
Specifications.No Class 1,2,3 or other solvents are used or produced in
the manufacturing or purification of the product. Filtered through a 0.2
micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR