Griffin Beakers, Polyproplyne, Graduated, Chemglass
Supplier: Chemglass
CG-1108-15 CG-1108-10 CG-1108-11 CG-1108-13 CG-1108-16 CG-1108-17
89028-348EA 120.72 CAD
89028-348 89028-338 89028-340 89028-344 89028-350 89028-352
Griffin Beakers, Polyproplyne, Graduated, Chemglass
Beakers
Low form polypropylene beakers.
  • Steam autoclavable at 121 °C
  • Chemically resistant
  • Printed graduations show approximated volume
Sold in the package quantities as listed.