Bellows Adapters, PTFE, Chemglass
Supplier: Chemglass
CG-1004-21 CG-1004-22 CG-1004-20
89028-198EA 368.82 CAD
89028-198 89028-200 89028-196
Bellows Adapters, PTFE, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter is ideal for eliminating stress between standard joints.
  • Constructed completely from PTFE
  • Can be bent up to 120°
  • Outer and inner joints are the same size