Murphy and Read Spring, Stainless Steel Springs, Chemglass
Supplier: Chemglass
CG-110-01 CG-110-03
80072-944PK 69.5 CAD
80072-944 80072-948
Murphy and Read Spring, Stainless Steel Springs, Chemglass
Springs
Stainless steel springs are used to connect apparatus with hooks.
  • Springs can easily connect to almost any size hook
  • Diameter of the coils on all sizes is 0.190"
  • Stainless steel construction
All springs have an enlarged 1/4" I.D. loop at each end for easy connection to almost any size hook. Length listed is the distance between loops.