Airfree® Schlenk Solvent Distillation Apparatus, Chemglass
Supplier: Chemglass
AF-0720-02 AF-0720-04
80060-168EA 3656.11 CAD
80060-168 80060-172
Airfree® Schlenk Solvent Distillation Apparatus, Chemglass
Distillation Components Distillation Sets
Apparatus designed for use in distillation of material such as THF or ether.
  • Storage reservoir has a capacity of 125 ml or 500 ml
  • 0 to 4 mm Chem-Vac™ Chem-Cap® valves ensure grease-free, leakproof seals
  • Top outer joint is a 24/40 while the uptake tube is a 24/40 inner joint
Extremely efficient cold finger coil condenser has a section that is immersed into the distillate. This assures that the distillate is at room temperature instead of the distillation temperature.
Side vapor tube (A) is fitted with a 14/20 standard taper outer joint, to be usedMore Product Information