Inner Luer Ground Joints, Chemglass
Supplier: Chemglass
CG-112-01 CG-112-02 CG-112-03
80072-958EA 36.8 CAD
80072-958 80072-960 80072-962
Inner Luer Ground Joints, Chemglass
Adapters Adapters, Joints with Tube Connectors
Inner joint manufactured to the same specifications found on medical and research syringes.
Complies with Federal Specification CG-S-921b and USAST Standard Z70.1. Available tooled on three tubing sizes with the inner diameter being 1 mm through the tip of the joint on all three sizes.