Inlet Adapters, with Hose Connection, Chemglass
Supplier: Chemglass
CG-1046-02 CG-1046-14
80066-030EA 154.92 CAD
80066-030 80066-032
Inlet Adapters, with Hose Connection, Chemglass
Adapters Adapters, Joints with Tube Connectors
With a serrated hose connection for vacuum or for the introduction of gases.
  • Equipped with a #7 Chem-Thread that creates a vacuum tight seal
  • Supplied complete with a compression cap and Viton O-Ring
  • For replacement compression cap, see CG-352-01 series
  • For replacement O-Ring, see CG-305-107 series
The #7 Chem-Thread at the top is for a vacuum tight seal of plain stem thermometers or any other tube having an O.D. between 4 to 7 mm. Serrated hose connection has an O.D. of 10 mm at the largest serration. Supplied complete with a compression cap and Viton O-Ring.