Universal Inlet Adapters, with Hose Connections, Chemglass
Supplier: Chemglass
CG-1045-01 CG-1045-04 CG-1045-05 CG-1045-06
80066-012EA 152.71 CAD
80066-012 80066-014 80066-016 80066-018
Universal Inlet Adapters, with Hose Connections, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapters have the listed size of Chem-Thread at the top for a vacuum tight seal on a variety of tubing diameters.
  • Will accept glass, metal, or heavy wall plastic tubes with diameters between 7 to 18 mm O.D.
  • Adapters supplied with a Celcon compression cap and Viton O-Ring
  • For replacement compression caps, see CG-352 series
  • For replacement O-Rings, see CG-305 series
Serrated hose connection has an O.D. of 10 mm at the largest serration. Adapters will accept glass, metal, or heavy wall plastic tubes such as thermometer wells, gas dispersion tubes, plainMore Product Information