Solvent Reservoir Bottles, Chemglass
Supplier: Chemglass
CG-1167-31 CG-1167-40 CG-1167-32 CG-1167-33 CG-1167-34
10170-268EA 374.3 CAD
10170-268 10170-498 10170-652 10170-696 10171-136
Solvent Reservoir Bottles, Chemglass
Bottles Solution Bottles
Solvent reservoir bottles, glass bottle only.
  • Plastic-coated flat bottom
  • Blue tinted
Bottles are blue tinted, with a plastic-coated flat bottom. The coating provides additional safety by containing the glass fragments from accidents and allowing enough time to transfer samples or discarding safely. For complete assembled solvent reservoir systems, see CG-1167-11, CG-1167-12, CG-1167-12, or CG-1167-14 series.