Aspirator Bottles, Graduated, Chemglass
Supplier: Chemglass
CG-1124-03 CG-1124-04 CG-1124-05 CG-1124-01 CG-1124-02
80066-366EA 435.85 CAD
80066-366 80066-368 80066-370 80066-362 80066-364
Aspirator Bottles, Graduated, Chemglass
Bottles Aspirators
Aspirator bottles feature permanent white graduations.
  • Reinforced tooled top accepts the listed size of rubber stopper
  • Use as a reservoir for fermentation work or as a large scale animal feeder
  • 250 and 500 ml bottles are graduated in 50 ml subdivisions
  • 1000, 2000, and 4000 ml bottles are graduated in 100 ml subdivisions
Hose connection has an Ext. Ø of 9 mm at the largest serration.
Order Now
SPECIFICATIONS
Environmentally Preferable