Connecting Adapters, Luer Outer Top and [ST] Inner Joint, Chemglass
Supplier: Chemglass
CG-1055-A-01 CG-1055-A-14
89028-274EA 110.13 CAD
89028-274 89028-278
Connecting Adapters, Luer Outer Top and [ST] Inner Joint, Chemglass
Adapters Adapters, Conical Ground Joints
Adapter has a luer outer on top and a lower standard taper inner joint.