Inlet Adapters, [ST] Inner Joint, Chemglass
Supplier: Chemglass
CG-1012-01 CG-1012-02 CG-1012-14 CG-1012-19
80065-678EA 100.56 CAD
80065-678 80065-680 80065-688 80065-690
Inlet Adapters, [ST] Inner Joint, Chemglass
Adapters Adapters, Conical Ground Joints
Allows for flexible tubing to be connected to apparatus.
  • Used for introduction of vacuum or pressure
  • Hose connection has an O.D. of 10 mm
  • Manufactured by Chemglass
Hose connection has an O.D. of 10 mm at the largest serration.