Heating Mantles, Upper Portion for Unjacketed Cylindrical Vessels, Chemglass
Supplier: Chemglass
CG-10013-10 CG-10013-15 CG-10013-20 CG-10013-21 CG-10013-30 CG-10013-50
16002-574EA 5296.92 CAD
16002-574 16002-576 16002-578 16002-580 16002-582 16002-584
Heating Mantles, Upper Portion for Unjacketed Cylindrical Vessels, Chemglass
Heaters Heating Mantles
Glass fabric heating mantle with a lengthwise slit, held together with a hook and lace closure.
  • For use with CG-1924 and CG-1972 unjacketed cylindrical reaction vessels
  • 115 volts
Delivery information: Supplied complete with a separable 4', 2-wire cord and locking connector.