Cryobox for PolarSafe™ Cryogenic Storage Vials with Star Caps, Argos Technologies
Supplier: Antyila Scientific
04395-84 04395-86 04395-87
10146-800EA 80.56 CAD
10146-800 10146-828 10146-830 10146-832
Cryobox for PolarSafe™ Cryogenic Storage Vials with Star Caps, Argos Technologies
Boxes Cryoboxes
The Argos PolarSafe polycarbonate storage boxes work in conjuciton with the 2D tubes to provide automatic identification of both the box and stored vials.
  • Each box features a 2D Datamatrix code on the bottom of the box as well as a laser engraved Code 128 barcode on the side Offered in both 81 and 100-well formats
PolarSafe 2D boxes are available in “short” (for 2 ml tubes) and “tall” designs (for 5 ml tubes).