Airfree® Schlenk Filter Tubes, Chemglass
Supplier: Chemglass
AF-0546-02 AF-0546-07 AF-0546-01 AF-0546-04 AF-0546-05 AF-0546-08
80064-874EA 798.08 CAD
80064-874 80064-884 80064-872 80064-878 80064-880 80064-886
Airfree® Schlenk Filter Tubes, Chemglass
Filters Filter Tubes
Similar to the AF-0544 series, but with the addition of two side arms.
  • Equipped with 2 mm high vacuum glass stopcocks
  • Availble in either coarse or medium frit porosity
  • Manufactured by Chemglass
The 2 mm high vacuum glass stopcocks have stainless steel constant pressure retaining devices.