Flow Control Adapters, 90°, with PTFE Stopcock, Chemglass
Supplier: Chemglass
CG-1030-45 CG-1030-19 CG-1030-29
16002-604EA 399.25 CAD
16002-604 80065-910 80065-912
Flow Control Adapters, 90°, with PTFE Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
Used to regulate the flow of gases or liquids into reaction systems.
  • Equipped with a PTFE stopcock
  • For replacement stopcocks, see CG-424-77
  • 90° angled
Hose connection has an O.D. of 8 mm at the middle serration with the exception of CG-1030-45, which has an O.D. of 14 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable