Flow Control Adapters, Straight, with Glass Stopcock, Chemglass
Supplier: Chemglass
CG-1029-G-01 CG-1029-G-02 CG-1029-G-14 CG-1029-G-19
80065-894EA 228.1 CAD
80065-894 80065-896 89028-206 89028-208
Flow Control Adapters, Straight, with Glass Stopcock, Chemglass
Adapters Adapters, Joints with Tube Connectors
Used to regulate the flow of gases or liquids into reaction systems.
  • Supplied with a CG-423 retaining clip
  • Equipped with 2mm glass stopcock
  • For replacement stopcocks, see CG-400-98
Stopcock has a bore of 2 mm and is supplied complete with a CG-423 retaining clip. Hose connection has an O.D. of 8 mm at the middle serration.