Distillation Adapters, with Two [ST] Joints, Chemglass
Supplier: Chemglass
CG-1072-01 CG-1072-14
80066-168EA 241.75 CAD
80066-168 80066-170
Distillation Adapters, with Two [ST] Joints, Chemglass
Distillation Components Distillation Adapters
With two standard taper inner joints, parallel, and at a 75° angle to the connecting tube.