Distillation Adapters, with Thermometer Joint, Chemglass
Supplier: Chemglass
CG-1024-02 CG-1024-14 CG-1024-01 CG-1024-19
80065-834EA 231.6 CAD
80065-834 80065-836 80065-832 80065-838
Distillation Adapters, with Thermometer Joint, Chemglass
Distillation Components Distillation Adapters
Adapters have a standard taper outer joint at the top of the main tube for use with jointed thermometers.
  • With inner joints on the bottom and sidearm at a 75° angle to one another
  • For use with the corresponding size jointed thermometers
  • Manufactured by Chemglass
The 10/18 size is used with 10/18, 25 mm immersion thermometers while the 10/30 is for use with 10/30, 75 mm immersion thermometers.