Seals for 8 mm Crimp-Top Vials, Thermo Scientific
Supplier: Thermo Scientific
C4008-2A C4008-1A C4008-4A
66030-822PK 292.68 CAD
66030-822 66011-180 66030-824
Seals for 8 mm Crimp-Top Vials, Thermo Scientific
Closures Chromatography Vial Closures
National Scientific aluminum crimp closures provide a secure leak-resistant seal.
  • Aluminium crimp seals with prefitted septa
  • Pre-assembled caps and septa are convenient and minimize contamination from handling
Available with a variety of septa materials with and without liners to match your chromatography needs. Choose from Thermo Scientific™, Thermo Scientific™ National™, and Thermo Scientific™ Chromacol™ brands.