Vacuum Filtration Adapters, with Pluro Stopper Tooled Tops, Chemglass
Supplier: Chemglass
CG-1052-01 CG-1052-02 CG-1052-03 CG-1052-04 CG-1052-14
80066-074EA 212.84 CAD
80066-074 80066-076 80066-078 80066-080 80066-082 80066-084
Vacuum Filtration Adapters, with Pluro Stopper Tooled Tops, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters are used for reduced pressure filtration with plain stem buchner funnels.
  • Top is tooled to accept the listed size of Pluro stopper
  • For replacement Pluro stoppers, See CG-1401 series
  • Manufactured by Chemglass
Serrated hose connection has an O.D. of 10 mm at the largest serration.