Universal Inlet Adapters, PTFE, Chemglass
Supplier: Chemglass
CG-1047-01 CG-1047-03 CG-1047-05 CG-1047-07 CG-1047-10 CG-1047-11 CG-1047-12 CG-1047-13 CG-1047-14 CG-1047-20
80060-282EA 211.75 CAD
80060-282 80060-286 80060-290 80060-294 80060-296 80060-298 80060-300 80060-302 80060-304 80060-306
Universal Inlet Adapters, PTFE, Chemglass
Adapters Adapters, Conical Ground Joints
Unique adapter is made entirely of PTFE and is supplied complete with a series of five PTFE ferrule-style cones.
  • Inside diameters run from 3.5-6.5 mm
  • Color-coded cones for easy identification
  • Replacement PTFE cones are available in sets of 5 or individually
These cones have inside diameters that run from 3.5 to 6.5 mm and seal against the inserted tube by means of a threaded compression screw. Cones are color-coded for easy identification.