Glas-Col® Series O Heating Mantles, for Cylindrical Vessels with Bottom Drain Openings, Chemglass
Supplier: Chemglass
CG-10009-10 CG-10009-12 CG-10009-14 CG-10009-16 CG-10009-18 CG-10009-20
80060-234EA 2137.02 CAD
80060-234 80060-236 80060-238 80060-240 80060-242 80060-244
Glas-Col® Series O Heating Mantles, for Cylindrical Vessels with Bottom Drain Openings, Chemglass
Heaters Heating Mantles
These Glas-Col® Series O mantles are similar to the CG-10007 series. However this mantle is slit lengthwise to a 1.5" hole in the bottom center with lacing closure to accommodate a cylindrical reaction vessel (CG-1924) with a bottom drain.
  • Mantle is glass fabric
  • 115 volts
  • Supplied with a separable 4', 2-wire extension cord with locking connector for each circuit
The 10,000 ml size has two circuits at the listed wattage.