Glas-Col® Heating Mantle Tops, Chemglass
Supplier: Chemglass
CG-10004-01 CG-10004-04 CG-10004-05 CG-10004-06 CG-10004-07 CG-10004-08 CG-10004-03
80060-200EA 733.71 CAD
80060-200 80060-208 80060-210 80060-212 80060-214 80060-216 80060-204
Glas-Col® Heating Mantle Tops, Chemglass
Heaters Heating Mantles
These Glas-Col® mantles help prevent vapor condensation in the upper half of a flask during distillation of high-boiling materials.
  • Heating Mantle Tops protect the flask, the solution, and the laboratory in multiple ways
  • Protect from stray drafts, which would disturb the reflux ratio during fractionation
  • Protect from flying glass of flask implosions in high-vacuum distillations
  • Protect from falling objects
  • Protect from thermal strain between top and lower half of flask during high-temperature operations
The top is designed for use with CG-11000, CG-12000, or CG-125000 mantles. With a glass-fabric construction and laced-side opening, an elliptical hole accommodates a 1, 2, and 3-neck flask. All mantles are 115 volts and have a 4', 2-wire cord with locking connector.