VWR® Moisture Analyzers
Supplier: VWR International
76289-740 76289-770 76289-772 76289-774 76290-332 76290-334
76289-740EA 6828.76 CAD
76289-740 76289-770 76289-772 76289-774 76290-332 76290-334
VWR® Moisture Analyzers
Analysers Moisture Analysers
The VWR® series Moisture Analyzers offer precise and reliable results for moisture content analysis.
  • Automatic opening and closing of drying chamber lid
  • Large, 5" color capacitive touch screen
  • Moisture content analysis
  • Dry mass determination
  • Hands-free operation with IR proximity sensors
  • Three-year warranty
The VWR® Moisture Analyzers can be utilized for a range of applications. Choose between an IR Emitter (max temperature 160 °C) or Halogen (max temperature 250 °C) heating module. For accurate moisture content analysis, the VWR® moisture analyzer features 4 drying profiles (standard, fast, step mild) and 4 finish modes (manual, automatic, time-defined, user-defined) to allow for timely, repeatable results.
The large, color capacitive touch screen allows for easy navigation of the VWR® moisture analyzers menus. The IR proximity sensors allow for hands free operation, easily program the sensors to perform actions such as opening/closing drying chamber lid, zeroing, taring, or starting an experiment.
Store up to 5000 products, 200 drying modes, 5000 drying reports, and 100 users within the VWR® Moisture Analyzers databases.
Certifications: C-UL(US) and CE.
Delivery information: Includes moisture analyzer and aluminum pan.