Headspace Caps, MicroSolv
Supplier: MicroSolv Technology Corp
95020-45S-CD 95020-45S-CD-K 95020-45S-T 95020-45S-TK 95020-45S-W 95020-45S-W-K 95020-55S-CD 95020-55S-CD-K 95020-55S-W 95020-55S-W-K 95025-01-1S 95025-01-2S 95025-02-1S 95025-02-2S 95025-04-1S 95025-04-2S 95025-04-3D 95025-04-4D 95025-04-5R 95025-07-1S 95025-08-1S 95025-08-2S 95025-09-1S 95025-14-1S 95025-01-3D 95020-21S-T 95020-40T-SK 95025-03-1S 95020-40T-S 95025-03-2S 95020-25S-T 95025-00-1S 95025-14-3D 95025-03-3D 95025-23-4D 95025-26-1D 95025-26-4B 95025-26-4R 95020-26-S 95020-50S-W 95020-50S-W-K
97054-910PK 435.73 CAD
97054-910 97054-912 97054-914 97054-916 97054-918 97054-920 97054-922 97054-924 97054-926 97054-928 97054-930 97054-932 97054-934 97054-936 97054-938 97054-940 97054-942 97054-944 97054-946 97054-948 97054-952 97054-954 97054-956 97054-962 97054-972 97054-974 97057-148 97057-164 97057-176 97057-232 97057-242 97057-244 97057-728 10014-046 10014-048 10014-050 10014-062 75997-576 75997-578 75997-580 75997-582 76057-644 76057-646
Headspace Caps, MicroSolv
Closures Chromatography Vial Closures
MicroSolv™ headspace caps are available as magnetic and with 4 different septa types to meet needs.
The screw cap eliminates the need for heavy crimping tools and assures that the vials are sealed perfectly every time. The design saves time and money, and ensures a good head space analysis.
Order Now
SPECIFICATIONS
Environmentally Preferable