Hybridization Bottles, Chemglass
Supplier: Chemglass
CG-1140-08 CG-1140-01 CG-1140-02 CG-1140-03 CG-1140-04 CG-1140-05 CG-1140-07 CG-1140-10 CG-1140-11 CG-1140-12 CG-1140-13 CG-1140-14 CG-1140-16 CG-1140-17
80066-422EA 378.97 CAD
80066-422 80066-410 80066-412 80066-414 80066-416 80066-418 80066-420 80066-424 80066-426 80066-428 80066-430 80066-432 80066-434 80066-436
Hybridization Bottles, Chemglass
Bottles Hybridisation Bottles/Tubes
Heavy-wall bottles are used in hybridization incubators or ovens with rotators.
  • Bottle fits most hybridization equipment
  • Straight interior walls for easy membrane placement and removal
  • Dual chamber bottles have a capped opening at each end with a solid glass wall in the center to create two equal compartments
  • Red cap has a PTFE liner and working temperature of 180 °C
  • Blue cap has silicone sealing ring and a working temperature of 140 °C
An alternative to plastic bags or boxes, the membrane is rolled and placed into the bottle which is secured in a rotisserie. The membrane is saturated and drained during each revolution, resulting in even exposure of the blots to probe or wash solutions. Bottle fits most hybridization equipment and has straight interior walls for easy membrane placement and removal, as well as decanting of hybridization solutions.