Inserts for Standard Opening Crimp-Top Vial, 11 mm, Thermo Scientific
Supplier: Thermo Scientific
C4012-529L C4012-529
66030-910PK 227.77 CAD
66030-910 66064-264 66065-190 66065-192 66030-908
Inserts for Standard Opening Crimp-Top Vial, 11 mm, Thermo Scientific
Vials
These inserts are made of clear, borosilicate glass.
These inserts are custom fitted for use with standard opening, 2 ml, 12×32 mm vials. Flat-bottom construction optimizes the depth of the vial.