Connecting/Reducing Adapters, Chemglass
Supplier: Chemglass
CG-1000-07 CG-1000-09 CG-1000-33 CG-1000-34 CG-1000-37 CG-1000-39 CG-1000-42 CG-1000-46
80065-478EA 122.12 CAD
80065-478 80065-482 80065-506 80065-508 80065-512 80065-516 80065-520 80065-528
Connecting/Reducing Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters are used to connect dissimilar standard taper joint sizes.
  • Lower inner joint is larger in size than the top outer joint
  • Adapters with joint combinations other than those listed are also available