Glas-Col® Stirmantle Dual Control, Ace Glass Incorporated
Supplier: Ace Glass
12048-02 12048-03
76291-802EA 3831.54 CAD
76291-802 76291-804
Glas-Col® Stirmantle Dual Control, Ace Glass Incorporated
Controllers Temperature Controllers
Combination stirring and temperature controller for Glas-Col stirmantles is often used to simultaneously power the Stirmantle's heating and stirring, as well as, an additional top mantle for those applications where vapor condensation on the upper half of the flask is undesirable.
  • Two-outlet version allows up to 1800 W
  • Three-outlet version allows up to 2400 W
Dual control features a universal J, T, and K probe input, a thermocouple break alarm, NEMA 5 - 15R receptacles, and a lab frame support bracket.
Order Now
SPECIFICATIONS
Environmentally Preferable