Glas-Col® Stir Mantles, Large Capacity, Ace Glass
Supplier: Ace Glass
12045-13 12045-23 12045-02 12045-03 12045-06
76291-790EA 5290.84 CAD
76291-790 76291-792 76291-870 76291-872 76291-874
Glas-Col® Stir Mantles, Large Capacity, Ace Glass
Heaters Heating Mantles
Aluminum housed mantle with magnetic stirrer 1/50 hP motor built-in. These mantles are ideal for continuous distillations by providing controlled heating and stirring in a single unit.
  • Variable speed control
  • Powerful motor or magnet combination generates speed range of 100 to 1850 rpm
  • Sized for 2 to 72 l spherical flasks
  • Large capacity
Independent speed and heat control is accomplished via the use of the Glas-Col® bundled speed control, combined with either another supplier's temperature control or a Glas-Col® combination control.
Order Now
SPECIFICATIONS
Environmentally Preferable