Economical Class 7 Slotted Weights
Supplier: Troemner
30390758 30390757 30390756 30390753 30390752 30390759 30390755 30390754 30390751 30390727 30390735 30390733 30390750
470013-400EA 54.1 CAD
470013-400 470013-402 470013-404 470013-406 470013-408 470017-692 470017-694 470017-696 470017-698 470121-072 470204-262 470204-264 470204-268
Economical Class 7 Slotted Weights
Weights Calibration Weights
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
  • Weights above 5 g are constructed of 303 Stainless Steel
  • Weights 5 g and below are constructed of 3003-H14 Aluminum
  • Weight Hanger sold separately
  • Manufactured in the USA
All weights are slotted and feature one-piece construction. Weight sets are supplied on a weight rack.