Economical Class 7 Cylindrical Weights
Supplier: Troemner
30390969 30390974 30390973 30390957 30391009 30391001 30391006 30391005 30391004 30391003 30391002 30391000 30390999 30390956 30391008 30391007
470120-998KT 165.9 CAD
470120-998 470204-252 470204-254 470204-258 470204-270 470204-282 470305-808 470305-810 470305-864 470305-866 470305-868 470305-870 470305-872 470305-874 470305-876 470305-878
Economical Class 7 Cylindrical Weights
Weights Calibration Weights
For rough weighing operations in physical and chemical laboratories, such as force measuring apparatus.
  • Weights above 5 g are constructed of 303 Stainless Steel
  • Weights 5 g and below are constructed of 3003-H14 Aluminum
  • Manufactured in the USA
These sets are supplied in a rugged plastic case with a locking latch and compartment for each weight in the case. Weights 100 g through 10 g are cylindrical with a handling knob. Weights 5 g through 1 g are flat with one end turned up for easy handling. Individual weights are not supplied in a case.