WarmStart LAMP Kit (DNA and RNA), New England Biolabs
Supplier: New England Biolabs (NEB)
E1700S E1700L
CA103258-588EA 359 CAD
CA103258-588 CA103258-590
WarmStart LAMP Kit (DNA and RNA), New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
Simple, one-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets.
  • Compatible with multiple detection methods.
  • WarmStart® technology inhibits activity at room temperature
The WarmStart LAMP Kit (DNA and RNA) is designed to provide a simple, one-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets. LAMP and RT-LAMP are commonly used isothermal amplification techniques that provides rapid detection of a target nucleic acid using LAMP-specific primers (supplied by the user) and a strand-displacing DNA polymerase.
WarmStart LAMP 2X Master Mix, which contains a blend of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase in an optimized LAMP buffer solution. Both Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase have been engineered for improved performance in LAMP and RT-LAMP reactions.
A fluorescent dye enables real-time fluorescence measurement of LAMP. The WarmStart LAMP Kit is compatible with multiple detection methods, including turbidity detection, real-time fluorescence detection (when used with LAMP fluorescent dye) and end-point visualization.
Delivery information: Supplied with WarmStart LAMP 2X Master Mix and a fluorescet dye.