PNGase A

Supplier: New England Biolabs (NEB)

P0707S P0707S P0707L
103257-616EA 502.44 CAD
103257-616 CA103257-616 CA103258-586
PNGase A
Enzymes

PNGase A cleaves N-glycans from plant and insect derived glycoproteins and glycopeptides.


  • Activity is not inhibited by an 1-3 Fucose residue on the chitobiose core
  • Recombinant enzyme with no detectable endoglycosidase or other exoglycosidase contaminating activity
  • Glycerol-free for optimal performance in HPLC and mass spectrometry analysis


PNGase A cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and short complex oligosaccharides such as those found in plant and insect cells from N-linked glycoproteins and glycopeptides. PNGase A differs from PNGase F in that it cleaves N-linked glycans with or without (1,3)-linked core fucose residues.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR