beta1-3,4 Galactosidase
Supplier: New England Biolabs (NEB)
P0746S
P0746S
103258-574EA
251.22
CAD
103258-574
CA103258-574
beta1-3,4 Galactosidase
Enzymes
Recombinant enzyme with no detectable endoglycosidase or other exoglycosidase contaminating activity.
- Glycerol-free for optimal performance in HPLC and mass spectrometry analysis
- ≥95% purity, as determined by SDS-PAGE and intact ESI-MS
- Optimal activity and stability for up to 12 months
β1-3,4 Galactosidase is an exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing β1-3 and β1-4-linked galactose residues from oligosaccharides.