BstZ17I-HF, Restriction Enzymes, New England Biolabs

Supplier: New England Biolabs (NEB)

R3594L R3594S
CA103258-570EA 480 CAD
CA103258-570 CA103258-572
BstZ17I-HF, Restriction Enzymes, New England Biolabs
Nucleic Acid Reagents Restriction Enzymes

An E. coli strain that carries the cloned and modified BstZ17I gene from Bacillus stearothermophilus 38M


  • Concentration: 20000 units/ml
  • 100% activity in CutSmart® buffer
  • Time-Saver™ qualified for digestion in 5 to 15 minutes
  • Reduced star activity


High-Fidelity (HF) restriction enzymes have the same specificity as native enzymes, but engineered for significantly reduced star activity and performance in a single buffer (CutSmart® Buffer).


Recombinant enzyme with no detectable exoglycosidase or other endoglycosidase contaminating activities.


Delivery information: All HF-restriction enzymes come with gel loading dye, purple (6X). Store at –20 °C.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR