Exo-Terra® Terraria
Supplier: ROLF C. HAGEN (USA) CORP.
PT2601 PT2605
470308-870KT 90.65 CAD
470308-870 470308-872
Exo-Terra® Terraria
Educational Materials Biology Educational Materials Plant Learning Activities
Easy sccess Terraria.
  • Front window ventilation
  • Raised bottom frame
  • Stainless steel mesh cover
  • Locks to prevent escape
  • Cut out in the back for wires and/or tubing
The Exo Terra Glass Terrarium is the ideal reptile or amphibian housing designed by European herpetologists. The front opening doors allow easy access for maintenance and feeding. A specially designed lock will prevent escape and the doors can be opened separately.
The full screen top ventilation allows UVB and infrared penetration, and is completely removable for easy access while decorating or cleaning. In the back of the screen cover are 5 closable wire or tube inlets on both sides to install Heat Wave Rocks, Waterfalls, sensors, etc. inside the terrarium. The bottom plate of the terrarium is raised to mount a substrate heater to heat a part of the extended ground surface.
The extra high fixed front window is ideal for thick layers of substrates (burrowing reptiles), aquatic parts (paludarium) or to mount an Exo Terra reptile den. The natural background is in the same color like all other Exo Terra decoration items (Water Dishes, Waterfalls, Heat Wave Rocks, etc.) and provides an extra climbing dimension.