VWR® 96-Well Deep Well Plates, PP
Supplier: VWR International
76210-518 76210-520 76210-522 76210-524
76210-518CS 629.98 CAD
76210-518 76210-520 76210-522 76210-524
VWR® 96-Well Deep Well Plates, PP
Microplates Deep Well Microplates
VWR® 96-Well Deep Well Plates are ideal for sample storage, sample re-suspension, cell culture, assay setup, diluting, and compound libraries. They are compatible with automated systems and are widely used in laboratories. Different sealing options are available, including cap seals or film seals, which can be applied independently to avoid cross-contamination.
  • DNase-free, RNase-free, and non-pyrogenic
  • Free from BSE and TSE
  • Latex-free
  • Sterilized by gamma irradiation to SAL 10–6 (ISO 11137)
  • 3-year shelf life following the month of production