Cole-Parmer® XRF Prep-Aid® Grinding Aids and Sample Binders
Supplier: SPEX SamplePrep
Cole-Parmer®
3644-150 3642-150 3642-450 3650 3644T-1500 3644-450 3644T-500 3646-150 3646-450
76533-164EA 1048.42 CAD
76533-164 76181-936 76182-408 76181-934 76182-410 76181-932 76182-412 76182-414 76533-152
Cole-Parmer® XRF Prep-Aid® Grinding Aids and Sample Binders
Mills
Prepare samples for reproducible and contamination-free results.
  • Select binders from powder, tablet, or liquid form depending on your application
High-quality sample preparation is the key to achieving consistent and reliable analytical results. Powders are ideal for samples that crumble or ablate after pressing. They blend quickly and completely with samples to form XRF (x-ray fluorescence) sample disks. Tablets are an efficient way to add binder to XRF sample disks. Liquid easily mixes with the sample.